3-Brom-2-thenoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromthiophen-2-carbonsäure, 3-Brom-2-thiophencarbonsäure

M-Clarity™ Qualitätsstufe = MQ200

633011-25G 633011-5G
SIAL633011-25GEA 111 EUR
SIAL633011-25G SIAL633011-5G
3-Brom-2-thenoesäure, Sigma-Aldrich®
3-Brom-2-thenoesäure
Formel: C₅H₃BrO₂S
Molecular Weight: 207,05 g/mol
MDL: MFCD00052291
CAS-Nummer: 7311-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR