5-Methyl-4-isoxazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

633771-10G 633771-1G
SIAL633771-10GEA 194 EUR
SIAL633771-10G SIAL633771-1G
5-Methyl-4-isoxazolcarbonsäure, Sigma-Aldrich®
5-Methylisoxazole-4-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127,1 g/mol
MDL: MFCD00955655
CAS-Nummer: 42831-50-5
EINECS: 222-289-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR