5-Isoxazolcarbonylchlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
636231-1G
636231-5G
SIAL636231-1GEA
80.5
EUR
SIAL636231-1G
SIAL636231-5G
5-Isoxazolcarbonylchlorid, Sigma-Aldrich®
5-Isoxazolcarbonylchlorid
Formel:
C₄H₂ClNO₂ Molecular Weight: 131,52 g/mol Siedepunkt: 74…76 °C (1013 hPa) |
MDL:
MFCD00067868 CAS-Nummer: 62348-13-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...