5-Amino-2-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Trifluormethyl)pyridin-5-amin, 2-(Trifluormethyl)-5-pyridylamin

M-Clarity™ Qualitätsstufe = MQ200

640107-1G 640107-5G
SIAL640107-1GEA 72.7 EUR
SIAL640107-1G SIAL640107-5G
5-Amino-2-(trifluormethyl)pyridin, Sigma-Aldrich®
5-Amino-2-(trifluormethyl)pyridin
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
MDL: MFCD01862656
CAS-Nummer: 106877-33-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR