1-Butyl-3-methylimidazoliumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Butyl-1-methyl-1H-imidazol-3-ium bromide

64133-25G 64133-5G
SIAL64133-25GEA 506 EUR
SIAL64133-25G SIAL64133-5G
1-Butyl-3-methylimidazoliumbromid, Sigma-Aldrich®
1-Butyl-3-methylimidazolium bromide
Formel: C₈H₁₅BrN₂
Molecular Weight: 219.12 g/mol
MDL: MFCD03427611
CAS-Nummer: 85100-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR