2-Chlor-4,6-difluoranilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

641413-1G 641413-5G
SIAL641413-1GEA 40.6 EUR
SIAL641413-1G SIAL641413-5G
2-Chlor-4,6-difluoranilin, Sigma-Aldrich®
2-Chlor-4,6-difluoranilin
Formel: C₆H₄ClF₂N
Molecular Weight: 163,55 g/mol
MDL: MFCD00142892
CAS-Nummer: 36556-56-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR