5-Bromthiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642517-1G 642517-5G
SIAL642517-1GEA 95.3 EUR
SIAL642517-1G SIAL642517-5G
5-Bromthiazol, Sigma-Aldrich®
5-Bromothiazole
Formel: C₃H₂BrNS
MDL: MFCD07787394
CAS-Nummer: 3034-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR