3-(2-Bromphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
643912-25G
643912-5G
SIAL643912-25GEA
144
EUR
SIAL643912-25G
SIAL643912-5G
3-(2-Bromphenyl)propionsäure, Sigma-Aldrich®
3-(2-Bromphenyl)propionsäure
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol |
MDL:
MFCD01310791 CAS-Nummer: 15115-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...