5-Methyl-3-isoxazolcarbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
644676-1G
644676-5G
SIAL644676-1GEA
27.8
EUR
SIAL644676-1G
SIAL644676-5G
5-Methyl-3-isoxazolcarbonsäure, Sigma-Aldrich®
5-Methylisoxazole-3-carboxylic acid
Formel:
C₅H₅NO₃ Molecular Weight: 127,1 g/mol |
MDL:
MFCD01318162 CAS-Nummer: 3405-77-4 EINECS: 222-289-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...