5-Amino-3-phenylisoxazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

644978-1G 644978-5G
SIAL644978-1GEA 61.4 EUR
SIAL644978-1G SIAL644978-5G
5-Amino-3-phenylisoxazol, Sigma-Aldrich®
5-Amino-3-phenylisoxazole
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD00464114
CAS-Nummer: 4369-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR