1-Methyl-3-phenylpiperazin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: MEFPIP

M-Clarity™ Qualitätsstufe = MQ100

648434-1G 648434-5G
SIAL648434-1GEA 33.9 EUR
SIAL648434-1G SIAL648434-5G
1-Methyl-3-phenylpiperazin, Sigma-Aldrich®
1-Methyl-3-phenylpiperazin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
MDL: MFCD03411603
CAS-Nummer: 5271-27-2
EINECS: 431-180-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR