2-Amino-5-chlorpyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlorpyrimidin-2-ylamin

M-Clarity™ Qualitätsstufe = MQ100

651567-1G 651567-5G
SIAL651567-1GEA 70.2 EUR
SIAL651567-1G SIAL651567-5G
2-Amino-5-chlorpyrimidin, Sigma-Aldrich®
2-Amino-5-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
MDL: MFCD00014604
CAS-Nummer: 5428-89-7
EINECS: 226-581-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR