1-(2,6-Difluorphenyl)-2-thioharnstoff, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2,6-Difluorphenylthioharnstoff, N-(2,6-Difluorphenyl)thioharnstoff
M-Clarity™ Qualitätsstufe = MQ100
654876-1G
654876-5G
SIAL654876-1GEA
53
EUR
SIAL654876-1G
SIAL654876-5G
1-(2,6-Difluorphenyl)-2-thioharnstoff, Sigma-Aldrich®
1-(2,6-Difluorphenyl)-2-thioharnstoff
Formel:
(C₆H₃F₂)NC(S)NH₂ Molecular Weight: 188,2 g/mol |
MDL:
MFCD05022144 CAS-Nummer: 59772-31-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...