3-Brom-4,5-diaminobenzotrifluorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Brom-5-(trifluormethyl)benzol-1,2-diamin, 1-Brom-2,3-diamino-5-(trifluormethyl)benzol, 3-Brom-5-(trifluormethyl)-1,2-phenylendiamin, 3-Brom-5-(trifluormethyl)-o-phenylendiamin, 3-Brom-4,5-diamino-α,α,α-trifluortoluol
M-Clarity™ Qualitätsstufe = MQ200
656003-1G
656003-5G
SIAL656003-1GEA
31.7
EUR
SIAL656003-1G
SIAL656003-5G
3-Brom-4,5-diaminobenzotrifluorid, Sigma-Aldrich®
3-Brom-4,5-diaminobenzotrifluorid
Formel:
C₇H₆BrF₃N₂ Molecular Weight: 255,04 g/mol |
MDL:
MFCD00042484 CAS-Nummer: 113170-72-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...