3-(4-Chlorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Chlorphenyl)propansäure

M-Clarity™ Qualitätsstufe = MQ100

656151-1G 656151-5G
SIAL656151-1GEA 32.5 EUR
SIAL656151-1G SIAL656151-5G
3-(4-Chlorphenyl)propionsäure, Sigma-Aldrich®
3-(4-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
MDL: MFCD00016555
CAS-Nummer: 2019-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR