3-(2,4-Dichlorphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(2,4-Dichlorphenyl)propansäure
M-Clarity™ Qualitätsstufe = MQ100
656186-1G
656186-5G
SIAL656186-1GEA
31.6
EUR
SIAL656186-1G
SIAL656186-5G
3-(2,4-Dichlorphenyl)propionsäure, Sigma-Aldrich®
3-(2,4-Dichlorphenyl)propionsäure
Formel:
C₉H₈Cl₂O₂ Molecular Weight: 219.07 g/mol |
MDL:
MFCD00275605 CAS-Nummer: 55144-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...