4-Amino-3-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

661090-100MG 661090-500MG
SIAL661090-100MGEA 26.1 EUR
SIAL661090-100MG SIAL661090-500MG
4-Amino-3-(trifluormethyl)pyridin, Sigma-Aldrich®
4-Amino-3-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
MDL: MFCD02183554
CAS-Nummer: 387824-61-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR