3-Methyl-1-hexin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
662887-10G
662887-1G
SIAL662887-10GEA
673
EUR
SIAL662887-10G
SIAL662887-1G
3-Methyl-1-hexin, Sigma-Aldrich®
3-Methyl-1-hexyne
Formel:
C₇H₁₂ Molecular Weight: 96,17 g/mol Siedepunkt: 85 °C (1013 hPa) Dichte: 0,719 g/cm³ (20 °C) |
CAS-Nummer:
40276-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...