3-Fluor-p-toluonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Fluoro-4-methylbenzonitrile
M-Clarity™ Qualitätsstufe = MQ100
664030-25G
664030-5G
SIAL664030-25GEA
130
EUR
SIAL664030-25G
SIAL664030-5G
3-Fluor-p-toluonitril, Sigma-Aldrich®
3-Fluor-p-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol |
MDL:
MFCD00153174 CAS-Nummer: 170572-49-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...