4-Fluorbenzoylacetonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(4-Fluorphenyl)-3-oxopropionsäurenitril, 3-(4-Fluorphenyl)-3-ketopropionsäurenitril
M-Clarity™ Qualitätsstufe = MQ100
681822-1G
681822-5G
SIAL681822-1GEA
42.7
EUR
SIAL681822-1G
SIAL681822-5G
4-Fluorbenzoylacetonitril, Sigma-Aldrich®
4-Fluorbenzoylacetonitril
Formel:
C₉H₆FNO Molecular Weight: 163,15 g/mol |
MDL:
MFCD00662062 CAS-Nummer: 4640-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...