3-Chlorbenzoylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(3-Chlorphenyl)-3-oxopropionsäurenitril, 3-(3-Chlorphenyl)-3-ketopropionsäurenitril

M-Clarity™ Qualitätsstufe = MQ100

681857-1G 681857-5G
SIAL681857-1GEA 32.6 EUR
SIAL681857-1G SIAL681857-5G
3-Chlorbenzoylacetonitril, Sigma-Aldrich®
3-Chlorbenzoylacetonitril
Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
MDL: MFCD00067891
CAS-Nummer: 21667-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR