4-Ethylbrenzcatechin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Ethylbenzol-1,2-diol

683957-1G 683957-5G
SIAL683957-1GEA 85.8 EUR
SIAL683957-1G SIAL683957-5G
4-Ethylbrenzcatechin, Sigma-Aldrich®
4-Ethylbrenzcatechin
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
MDL: MFCD00015847
CAS-Nummer: 1124-39-6
EINECS: 214-397-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR