4-(1-Aminoethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Pyridyl)ethylamin, 1-Pyridin-4-ylethylamin

M-Clarity™ Qualitätsstufe = MQ100

685747-1G 685747-250MG
SIAL685747-1GEA 95 EUR
SIAL685747-1G SIAL685747-250MG
4-(1-Aminoethyl)pyridin, Sigma-Aldrich®
4-(1-Aminoethyl)pyridine
Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
MDL: MFCD02245004
CAS-Nummer: 50392-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR