2-(Hydroxymethyl)indol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Indolcarbinol, (Indol-2-yl)methanol, Indol-2-carbinol, (2-Indolyl)methanol, Indol-2-methanol, 2-Indolmethanol
M-Clarity™ Qualitätsstufe = MQ100
687227-1G
687227-5G
SIAL687227-1GEA
70.2
EUR
SIAL687227-1G
SIAL687227-5G
2-(Hydroxymethyl)indol, Sigma-Aldrich®
2-(Hydroxymethyl)indole
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol |
MDL:
MFCD00030195 CAS-Nummer: 24621-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...