1-Decyl-3-methylimidazoliumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-decyl-3-methylimidazolium chloride
M-Clarity™ Qualitätsstufe = MQ100
690597-50G
690597-5G
SIAL690597-50GEA
375
EUR
SIAL690597-50G
SIAL690597-5G
1-Decyl-3-methylimidazoliumchlorid, Sigma-Aldrich®
1-Decyl-3-methylimidazolium chloride
Formel:
C₁₄H₂₇ClN₂ Molecular Weight: 258,83 g/mol |
MDL:
MFCD03427605 CAS-Nummer: 171058-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...