3-Fluor-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Cyan-3-fluorpyridin, 3-Fluor-2-pyridincarbonsäurenitril

M-Clarity™ Qualitätsstufe = MQ100

697109-1G 697109-5G
SIAL697109-1GEA 26 EUR
SIAL697109-1G SIAL697109-5G
3-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
3-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122,1 g/mol
MDL: MFCD06797501
CAS-Nummer: 97509-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR