5-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Cyan-5-fluorpyridin, 5-Fluor-2-pyridincarbonsäurenitril
M-Clarity™ Qualitätsstufe = MQ100
703982-1G
703982-5G
SIAL703982-1GEA
28.8
EUR
SIAL703982-1G
SIAL703982-5G
5-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
5-Fluor-2-pyridincarbonitril
Formel:
C₆H₃FN₂ Molecular Weight: 122,1 g/mol |
MDL:
MFCD08741371 CAS-Nummer: 327056-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...