6-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Fluor-2-pyridincarbonsäurenitril, 2-Cyan-6-fluorpyridin
M-Clarity™ Qualitätsstufe = MQ100
704024-1G
704024-250MG
SIAL704024-1GEA
89.1
EUR
SIAL704024-1G
SIAL704024-250MG
6-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
6-Fluor-2-pyridincarbonitril
Formel:
C₆H₃FN₂ Molecular Weight: 122,1 g/mol |
MDL:
MFCD09263323 CAS-Nummer: 3939-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...