1-(4-Pyridyl)piperazin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(1-Piperazinyl)pyridin
76763-1G-F
76763-5G-F
SIAL76763-1G-FEA
64
EUR
SIAL76763-1G-F
SIAL76763-5G-F
1-(4-Pyridyl)piperazin, Sigma-Aldrich®
1-(4-Pyridyl)piperazin
Formel:
C₉H₁₃N₃ Molecular Weight: 163,22 g/mol Siedepunkt: 195…200 °C (0,3 mmHg) Schmelzpunkt: 140 °C |
MDL:
MFCD00040745 CAS-Nummer: 1008-91-9 EINECS: 213-764-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...