m-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: m-Phenylendiammoniumdichlorid, 1,3-Phenylendiamin Dihydrochlorid, Benzol-1,3-diamindihydrochlorid, 1,3-Diaminobenzol Dihydrochlorid, 1,3-Phenylendiammoniumdichlorid

78450-100G 78450-500G
SIAL78450-100GEA 197 EUR
SIAL78450-100G SIAL78450-500G
m-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®
m-Phenylendiamin Dihydrochlorid
Formel: C₆H₁₀Cl₂N₂
Molecular Weight: 181,06 g/mol
Schmelzpunkt: 277…278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012975
CAS-Nummer: 541-69-5
EINECS: 208-790-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR