3,3',5,5'-Tetramethylbenzidin (TMB) ≥98%, Sigma-Aldrich®
Lieferant: Merck
Synonyme: TMB
M-Clarity™ Qualitätsstufe = MQ300
87748-1G 87748-25G 87748-5G
SIAL87748-1GEA 108 EUR
SIAL87748-1G SIAL87748-25G SIAL87748-5G
3,3',5,5'-Tetramethylbenzidin (TMB) ≥98%, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin (TMB)
3,3′,5,5′-Tetramethylbenzidine (TMB) is a noncarcinogenic substitute (Ames test negative) for benzidine suitable as peroxidase substrate for use in ELISA procedures.
The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
Formel: C₁₆H₂₀N₂
Molecular Weight: 240,35 g/mol
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007748
CAS-Nummer: 54827-17-7
EINECS: 259-364-6