1-Butyl-3-methylimidazoliumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Butyl-1-methyl-1H-imidazol-3-ium bromide

M-Clarity™ Qualitätsstufe = MQ100

95137-50G-F 95137-5G-F
SIAL95137-50G-FEA 136 EUR
SIAL95137-50G-F SIAL95137-5G-F
1-Butyl-3-methylimidazoliumbromid, Sigma-Aldrich®
1-Butyl-3-methylimidazolium bromide
Formel: C₈H₁₅BrN₂
Molecular Weight: 219,12 g/mol
MDL: MFCD03427611
CAS-Nummer: 85100-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR