4,5-Dimethyl-o-phenylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Diamino-4,5-dimethylbenzol, 4,5-Diamino-1,2-dimethylbenzol, 4,5-Dimethylbenzol-1,2-diamin, 4,5-Dimethyl-1,2-phenylendiamin

M-Clarity™ Qualitätsstufe = MQ100

D176605-5G D176605-25G
SIALD176605-5GEA 99.6 EUR
SIALD176605-5G SIALD176605-25G
4,5-Dimethyl-o-phenylendiamin, Sigma-Aldrich®
4,5-Dimethyl-o-phenylendiamin
Formel: C₈H₁₂N₂
MDL: MFCD00007729
CAS-Nummer: 3171-45-7
EINECS: 221-635-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR