Gemfibrozil, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeriansäure, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentansäure
M-Clarity™ Qualitätsstufe = MQ100
G9518-5G
G9518-25G
SIALG9518-5GEA
81
EUR
SIALG9518-5G
SIALG9518-25G
Gemfibrozil, Sigma-Aldrich®
Gemfibrozil
Formel:
C₁₅H₂₂O₃ Molecular Weight: 250.34 g/mol Siedepunkt: 395 Schmelzpunkt: 59…61 °C |
MDL:
MFCD00079335 CAS-Nummer: 25812-30-0 EINECS: 247-280-2 Merck Index: 12,04394 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...