3-Hydroxy-5-methylisoxazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hymexazol

H3275-250MG H3275-50MG
SIALH3275-250MGEA 441 EUR
SIALH3275-250MG SIALH3275-50MG
3-Hydroxy-5-methylisoxazol, Sigma-Aldrich®
3-Hydroxy-5-methylisoxazole
Formel: C₄H₅NO₂
Molecular Weight: 99,09 g/mol
MDL: MFCD00144468
CAS-Nummer: 10004-44-1
EINECS: 233-000-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR