1-Methyl-2-thioharnstoff, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
M84607-100G
M84607-25G
SIALM84607-100GEA
114
EUR
SIALM84607-100G
SIALM84607-25G
1-Methyl-2-thioharnstoff, Sigma-Aldrich®
1-Methyl-2-thioharnstoff
Formel:
C₂H₆N₂S Molecular Weight: 90,15 g/mol Schmelzpunkt: 119 °C |
MDL:
MFCD00004938 CAS-Nummer: 598-52-7 EINECS: 209-936-6 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...