Kaliumtellurit ≥90%, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
di-Kaliumtellurtrioxid, Kaliumtellur(IV)oxid
P0677-100G
P0677-25G
SIALP0677-100GEA
433
EUR
SIALP0677-100G
SIALP0677-25G
Kaliumtellurit ≥90%, Sigma-Aldrich®
Kaliumtellurit
Formel:
K₂TeO₃ Molecular Weight: 253,79 g/mol |
MDL:
MFCD00011379 CAS-Nummer: 7790-58-1 EINECS: 232-213-1 UN: 3284 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...