o-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: o-Phenylendiammoniumdichlorid

M-Clarity™ Qualitätsstufe = MQ300

P1526-100G P1526-10G P1526-25G P1526-50G
SIALP1526-100GEA 387 EUR
SIALP1526-100G SIALP1526-10G SIALP1526-25G SIALP1526-50G
o-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®
o-Phenylendiamin Dihydrochlorid
Formel: C₆H₁₀Cl₂N₂
Molecular Weight: 181,06 g/mol
Schmelzpunkt: 205 °C
MDL: MFCD00012966
CAS-Nummer: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR