2-Thenoesäurehydrazid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid
M-Clarity™ Qualitätsstufe = MQ200
T1388-1G
T1388-25G
T1388-5G
SIALT1388-1GEA
29.8
EUR
SIALT1388-1G
SIALT1388-25G
SIALT1388-5G
2-Thenoesäurehydrazid, Sigma-Aldrich®
2-Thenoesäurehydrazid
Formel:
C₅H₆N₂OS Molecular Weight: 142,18 g/mol Schmelzpunkt: 142…143 °C |
MDL:
MFCD00005435 CAS-Nummer: 2361-27-5 EINECS: 219-107-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...