Trimethylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Trimethylamin Hydrochlorid
M-Clarity™ Qualitätsstufe = MQ200
T72761-100G
T72761-25G
T72761-500G
SIALT72761-100GEA
30.2
EUR
SIALT72761-100G
SIALT72761-25G
SIALT72761-500G
Trimethylammoniumchlorid, Sigma-Aldrich®
Trimethylammonium chloride
Formel:
C₃H₁₀ClN Molecular Weight: 95.57 g/mol Schmelzpunkt: 273…278 °C Dichte: ∼1 g/cm³ (20 °C) |
MDL:
MFCD00012478 CAS-Nummer: 593-81-7 EINECS: 209-810-0 Merck Index: 12,09841 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...