2,6-Dimethyl-5-heptenal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethylhept-5-enal

M-Clarity™ Qualitätsstufe = MQ300

W238902-100G-K W238902-1KG-K
SIALW238902-100G-KEA 81.2 EUR
SIALW238902-100G-K SIALW238902-1KG-K
2,6-Dimethyl-5-heptenal, Sigma-Aldrich®
2,6-Dimethyl-5-heptenal
Formel: C₉H₁₆O
MDL: MFCD00006981
CAS-Nummer: 106-72-9
EINECS: 203-427-2
UN: 1989
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR