Ethylisovalerat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

M-Clarity™ Qualitätsstufe = MQ100

W246328-100G-K W246328-1KG-K
SIALW246328-100G-KEA 85.3 EUR
SIALW246328-100G-K SIALW246328-1KG-K
Ethylisovalerat, Sigma-Aldrich®
Ethylisovalerat
Formel: C₇H₁₄O₂
Molecular Weight: 130,19 g/mol
Siedepunkt: 131…133 °C (1013 hPa)
Schmelzpunkt: –99 °C
Dichte: 0,86 g/cm³ (20 °C)
MDL: MFCD00009203
CAS-Nummer: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 13,03849

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR