Methylisovalerat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isopropyl acetic acid methyl ester, Isovaleric acid methyl ester, 3-Methylbutyric acid emthyl ester, Methyl isopentanoate, 3-Methylbutanoic acid methyl ester

M-Clarity™ Qualitätsstufe = MQ300

W275301-1KG-K W275301-4KG-K
SIALW275301-1KG-KEA 103 EUR
SIALW275301-1KG-K SIALW275301-4KG-K
Methylisovalerat, Sigma-Aldrich®
Methylisovalerat
Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
Siedepunkt: 114 °C (1013 hPa)
Dichte: 0,90065 g/cm³ (0 °C)
MDL: MFCD00042866
CAS-Nummer: 556-24-1
EINECS: 209-117-3
UN: 2400
ADR: 3,II
Merck Index: 12,06166

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR