3-(Methylthio)propylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Methylthio)propylisothiocyanate

M-Clarity™ Qualitätsstufe = MQ300

W331201-100G-K W331201-1KG-K W331201-25G-K
SIALW331201-100G-KEA 180 EUR
SIALW331201-100G-K SIALW331201-1KG-K SIALW331201-25G-K
3-(Methylthio)propylisothiocyanat, Sigma-Aldrich®
3-(Methylthio)propylisothiocyanat
Formel: C₅H₉NS₂
Molecular Weight: 147.27 g/mol
MDL: MFCD00209521
CAS-Nummer: 505-79-3
EINECS: 208-020-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR