(2Z)-6-Nonen-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Z)-6-Nonen-1-ol

M-Clarity™ Qualitätsstufe = MQ300

W346500-100G-K W346500-1KG-K
SIALW346500-100G-KEA 367 EUR
SIALW346500-100G-K SIALW346500-1KG-K
(2Z)-6-Nonen-1-ol, Sigma-Aldrich®
cis-6-Nonen-1-ol
Formel: C₉H₁₈O
MDL: MFCD00015388
CAS-Nummer: 35854-86-5
EINECS: 252-764-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR