(2Z)-2-Nonen-1-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(Z)-2-Nonen-1-ol
M-Clarity™ Qualitätsstufe = MQ300
W372005-100G-K
W372005-25G-K
SIALW372005-100G-KEA
532
EUR
SIALW372005-100G-K
SIALW372005-25G-K
(2Z)-2-Nonen-1-ol, Sigma-Aldrich®
cis-2-Nonen-1-ol
Formel:
C₉H₁₈O Molecular Weight: 142,24 g/mol |
MDL:
MFCD00063210 CAS-Nummer: 41453-56-9 EINECS: 255-376-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...