Allyltriphenylsilan
Lieferant: CHEMPUR SILANE
SILS00475.5
SILS00475.25
SILNSILS00475.5EA
0
EUR
SILNSILS00475.5
SILNSILS00475.25
Allyltriphenylsilan
Allyltriphenylsilan
Formel:
H₂CCHCH₂Si(C₆H₅)₃ Molecular Weight: 300,48 g/mol |
MDL:
MFCD00077644 CAS-Nummer: 18752-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...