Benzyltrimethylsilan
Lieferant: CHEMPUR SILANE
SILS01050.10
SILS01050.25
SILS01050.100
SILNSILS01050.10EA
0
EUR
SILNSILS01050.10
SILNSILS01050.25
SILNSILS01050.100
Benzyltrimethylsilan
Benzyltrimethylsilan
Formel:
C₁₀H₁₆Si Molecular Weight: 164.32 g/mol Siedepunkt: 37…40 °C (0.8 mmHg) Dichte: 0.872 g/cm³ (15 °C) |
MDL:
MFCD00039796 CAS-Nummer: 770-09-2 EINECS: 212-218-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...