Bis(diethylamino)dimethylsilan
Lieferant: CHEMPUR SILANE
Synonyme:
N,N,N',N'-Tetraethyl-1,1-dimethylsilanediamine
SILS01550.10
SILS01550.25
SILS01550.50
SILNSILS01550.10EA
0
EUR
SILNSILS01550.10
SILNSILS01550.25
SILNSILS01550.50
Bis(diethylamino)dimethylsilan
Bis(diethylamino)dimethylsilan
Formel:
C₁₀H₂₆N₂Si Molecular Weight: 202,42 g/mol |
MDL:
MFCD00039879 CAS-Nummer: 4669-59-4 EINECS: 225-116-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...