Bis(dimethylamino)dimethylsilan

Lieferant: CHEMPUR SILANE

SILS01600.25 SILS01600.100
SILNSILS01600.25EA 0 EUR
SILNSILS01600.25 SILNSILS01600.100
Bis(dimethylamino)dimethylsilan
Bis(dimethylamino)dimethylsilan
Formel: C₆H₁₈N₂Si
Molecular Weight: 146,31 g/mol
Siedepunkt: 127,5 °C (1013 hPa)
Schmelzpunkt: –98 °C
Dichte: 0,809 g/cm³ (22 °C)
MDL: MFCD00008296
CAS-Nummer: 3768-58-9
EINECS: 223-200-1
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR